Learn More
Selleck Chemical LLC THZ1 2HCl
SDP
Supplier:  Selleck Chemical LLC S754925MG
Encompass_Preferred
THZ1 is a covalent CDK7 inhibitor which has the unprecedented ability to target a remote cysteine residue located outside of the canonical kinase domain providing an unanticipated means of achieving selectivity for CDK7
Catalog No. 50-000-04693
Ajouter au panier
missing translation for 'productTitle'
missing translation for 'pleaseSelectIssue'
missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.