Learn More
Selleck Chemical LLC THZ1 2HCl
SDP
Catalog No. 5000004547
Encompass_Preferred
THZ1 is a covalent CDK7 inhibitor which has the unprecedented ability to target a remote cysteine residue located outside of the canonical kinase domain providing an unanticipated means of achieving selectivity for CDK7
Catalog No. 50-000-04547 Supplier Selleck Chemical LLC Supplier No. S754910MM/1ML
THZ1 is a covalent CDK7 inhibitor which has the unprecedented ability to target a remote cysteine residue located outside of the canonical kinase domain providing an unanticipated means of achieving selectivity for CDK7