Learn More
TARGETMOL CHEMICALS INC THZ1 2HCL 5MG
SDP
Catalog No. 502580223
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. THZ1 2HCl (THZ1 Dihydrochloride) is a selective covalent and allosteric inhibitor of CDK7 with an IC50 of 3.2 nM. THZ1 2HCl has antiproliferative effects on a variety of cancer cell lines. purity: 98%
Catalog No. 50-258-0223 Supplier TARGETMOL CHEMICALS INC Supplier No. T353325MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. THZ1 2HCl (THZ1 Dihydrochloride) is a selective covalent and allosteric inhibitor of CDK7 with an IC50 of 3.2 nM. THZ1 2HCl has antiproliferative effects on a variety of cancer cell lines. purity: 98%