Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thz1 (hydrochloride) | 1604810-83-4 | MFCD28167785 | 99.1% | 602.51 g/mol | C31H29Cl2N7O2 | 10 MG
SDP

Catalog No. 5000298662
Encompass_Preferred

THZ1 (hydrochloride) is the hydrochloride salt of THZ1, a selective, covalent inhibitor of cyclin-dependent kinase 7 (CDK7) with low-nanomolar potency. It also inhibits CDK12 and CDK13 and is used in preclinical studies to probe transcriptional regulation, cell-cycle control, and oncology models.

  • Selective covalent CDK7 inhibitor (IC50 ~3.2 nM).
  • Hydrochloride salt form for improved handling.
  • Molecular formula C31H29Cl2N7O2; molecular weight 602.51 g/mol.
  • High purity (~99.06%) suitable for research applications.
  • Supplied in small research sizes, including 10 MG.
  • Certificate of analysis and safety data sheet available.
  • Common applications: transcriptional regulation, cell-cycle, and oncology research.

Catalog No. 50-002-98662 Supplier Medchemexpress LLC Supplier No. HY80013A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

THZ1 (hydrochloride) is the hydrochloride salt of THZ1, a selective, covalent inhibitor of cyclin-dependent kinase 7 (CDK7) with low-nanomolar potency. It also inhibits CDK12 and CDK13 and is used in preclinical studies to probe transcriptional regulation, cell-cycle control, and oncology models.

  • Selective covalent CDK7 inhibitor (IC50 ~3.2 nM).
  • Hydrochloride salt form for improved handling.
  • Molecular formula C31H29Cl2N7O2; molecular weight 602.51 g/mol.
  • High purity (~99.06%) suitable for research applications.
  • Supplied in small research sizes, including 10 MG.
  • Certificate of analysis and safety data sheet available.
  • Common applications: transcriptional regulation, cell-cycle, and oncology research.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.