Learn More
Medchemexpress LLC Thz1 (hydrochloride) | 99.1% | C31H29Cl2N7O2 | 10MM/1ML
SDP
Catalog No. 5000432753
Encompass_Preferred
THZ1 Hydrochloride is a selective and potent covalent inhibitor of CDK7, with an IC50 of 3.2 nM. It also inhibits the closely related kinases CDK12 and CDK13, leading to the downregulation of MYC expression.
  • Functions as a selective and potent covalent CDK7 inhibitor.
  • Inhibits closely related kinases, specifically CDK12 and CDK13.
  • Downregulates MYC expression.
Catalog No. 50-004-32753 Supplier Medchemexpress LLC Supplier No. HY80013A10MM/1ML
THZ1 Hydrochloride is a selective and potent covalent inhibitor of CDK7, with an IC50 of 3.2 nM. It also inhibits the closely related kinases CDK12 and CDK13, leading to the downregulation of MYC expression.
  • Functions as a selective and potent covalent CDK7 inhibitor.
  • Inhibits closely related kinases, specifically CDK12 and CDK13.
  • Downregulates MYC expression.