Learn More
Apexbio Technology LLC THZ2 1604810-84-5 10mM (in 1mL DMSO)
SDP
Catalog No. 5000061776
Encompass_Preferred
THZ2 (CAS 1604810-84-5) is a selective inhibitor of cyclin-dependent kinase 7 (CDK7) demonstrating an IC50 of 13 9 nM CDK7 a serine/threonine kinase bound and activated by cyclins regulates cell cycle progression and transcriptional control through phosphorylation of RNA polymerase II (RNAPII) In triple-negative breast cancer (TNBC) cell models THZ2 reduces cellular proliferation induces apoptotic cell death without affecting cell-cycle distribution and inhibits colony formation at low-nanomolar concentrations (IC50 10 nM) In xenograft mouse models THZ2 suppresses tumor growth without noticeable toxicity Thus THZ2 is useful in research investigating CDK7-mediated oncogenic mechanisms in cancer
Catalog No. 50-000-61776 Supplier Apexbio Technology LLC Supplier No. A87175.1
 Restricted Item
Add to Cart
Add to Cart
THZ2 (CAS 1604810-84-5) is a selective inhibitor of cyclin-dependent kinase 7 (CDK7) demonstrating an IC50 of 13 9 nM CDK7 a serine/threonine kinase bound and activated by cyclins regulates cell cycle progression and transcriptional control through phosphorylation of RNA polymerase II (RNAPII) In triple-negative breast cancer (TNBC) cell models THZ2 reduces cellular proliferation induces apoptotic cell death without affecting cell-cycle distribution and inhibits colony formation at low-nanomolar concentrations (IC50 10 nM) In xenograft mouse models THZ2 suppresses tumor growth without noticeable toxicity Thus THZ2 is useful in research investigating CDK7-mediated oncogenic mechanisms in cancer