Learn More
Medchemexpress LLC HY-103618 10mg Medchemexpress, THZ531 CAS:1702809-17-3 Purity:>98%
SDP
Catalog No. 502028735
Encompass_Preferred
Medchemexpress, HY-103618 10mg THZ531 CAS:1702809-17-3 THZ531 is a selective and covalent inhibitor of both CDK12 and CDK13 with IC50s of 158 nM and 69 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8735 Supplier Medchemexpress LLC Supplier No. HY10361810MG
Medchemexpress, HY-103618 10mg THZ531 CAS:1702809-17-3 THZ531 is a selective and covalent inhibitor of both CDK12 and CDK13 with IC50s of 158 nM and 69 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.