Learn More
IDEX Health & Science Upchurch TI FILTR END FITTING .5U 10/PK
Catalog No. NC1920950
Encompass_Preferred
Idex Inlet Solvent Filters: End Fittings, Pore Size: 0.5 um, Replacement Filter End Fitting, PEEK, Thread Size: 10 to 32 Coned
Catalog No. NC1920950 Supplier IDEX Health & Science Upchurch Supplier No. A428X
Idex Inlet Solvent Filters: End Fittings, Pore Size: 0.5 um, Replacement Filter End Fitting, PEEK, Thread Size: 10 to 32 Coned