Learn More
Biocare Medical, LLC TIA-1 Antibody - Concentrate, 0.1 ml
SDP
Supplier:  Biocare Medical, LLC CM130A
Encompass_Preferred
TIA-1 antibody (T-cell intracytoplasmic antigen) is expressed in lymphocytes processing cytolytic potential. Studies show that 60 to 70% of anaplastic large cell lymphoma reacts with TIA-1. Studies also indicate that TIA-1 reacts with most large granular lymphocytic leukemias, hepatosplenic T-cell lymphomas, intestinal T-cell lymphomas, NK-like T-cell lymphomas, NK cell lymphomas, nasal T/NK-cell lymphomas, subcutaneous T-cell lymphomas and pulmonary angiocentric lymphomas of T-or NK-phenotype. All B-cell lymphomas, Hodgkin and lymphoblastic leukemias were negative for TIA-1.
Catalog No. 50-825-39