Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 7072MSM1P1
Encompass_Preferred
Product Type: Antibody. Application: WB. Conjugate/Tag: Unconjugated. Clone: TIA1/1313. Clonality: Monoclonal. Host: Mouse. Isotype: IgG2b. Positive Control: HepG2, PC3 and HeLa cells. Tonsil. Cellular Staining: Cytoplasmic. Buffer: 10mM PBS with 0.05% BSA and 0.05% Azide. Concentration: 200 ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant human TIA1 fragment of 102 amino acid residues (aa279-380). Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 40kDa.Chromosome: 2p13. Gene Symbol: TIA1. Gene ID: 7072. Ensemble: ENSG00000116001. Uniprot: XP_016860286.
Catalog No. 50-188-5220