Learn More
AdipoGen Tiadinil
SDP
Supplier:  AdipoGen CDXT0121M100
Encompass_Preferred
Chemical. CAS 223580-51-6. Formula C11H10ClN3OS. MW 267.7. Synthetic. Systemic anilide fungicide. Mode of action is through induc tion of plant defense mechanisms against a pathogen, therefore being a host plant defense inducer. Compound can be used as analytical reference material.
Catalog No. 50-255-9994