Learn More
Medchemexpress LLC HY-137551 5mg Medchemexpress, Tiafenacil CAS:1220411-29-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1375515MG
Encompass_Preferred
Medchemexpress, HY-137551 5mg Tiafenacil CAS:1220411-29-9 Tiafenacil is a new protoporphyrinogen IX oxidase (PPO)-inhibiting herbicide, with IC50 values of 22 to 28 nM for various plant species, including amaranth (Amaranthus tuberculatus), soybean (Glycine max), arabidopsis (Arabidopsis thaliana), and rapeseed (Brassica napus). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1499