Learn More
Medchemexpress LLC HY-B0696A 10mg Medchemexpress, Tiagabine (hydrochloride) CAS:145821-59-6 Purity:>98%
SDP
Catalog No. 502082643
Encompass_Preferred
Medchemexpress, HY-B0696A 10mg Tiagabine (hydrochloride) CAS:145821-59-6 Tiagabine hydrochloride is a potent and selective GABA reuptake inhibitor, used as an anticonvulsant agent, with IC50s of 67, 446 and 182 nM for [3H]GABA uptake in Synaptosomes, Neurons and Glia, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2643 Supplier Medchemexpress LLC Supplier No. HYB0696A10MG
Add to Cart
Add to Cart
Medchemexpress, HY-B0696A 10mg Tiagabine (hydrochloride) CAS:145821-59-6 Tiagabine hydrochloride is a potent and selective GABA reuptake inhibitor, used as an anticonvulsant agent, with IC50s of 67, 446 and 182 nM for [3H]GABA uptake in Synaptosomes, Neurons and Glia, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.