Learn More
Selleck Chemical LLC Tiagabine hydrochloride S4661-100mg
SDP
Catalog No. 503111020
Encompass_Preferred
Tiagabine Hydrochloride (Gabitril NO050328 NO328 TGB) is the hydrochloride salt form of tiagabine a nipecotic acid derivative with anticonvulsant property A selective gamma-aminobutyric acid (GABA) reuptake inhibitor
Catalog No. 50-311-1020 Supplier Selleck Chemical LLC Supplier No. S4661100MG
Tiagabine Hydrochloride (Gabitril NO050328 NO328 TGB) is the hydrochloride salt form of tiagabine a nipecotic acid derivative with anticonvulsant property A selective gamma-aminobutyric acid (GABA) reuptake inhibitor