Learn More
Medchemexpress LLC Tiagabine (hydrochloride) | 145821-59-6 | MFCD00892911 | ≥98% | 1 ML
SDP
Catalog No. 5000407068
Encompass_Preferred
Tiagabine (hydrochloride) is an anticonvulsant agent and a selective inhibitor of the GAT-1 GABA transporter. It potently and selectively inhibits GABA reuptake and demonstrates neuroprotective properties.
  • Selective inhibitor of GAT-1 GABA transporter
  • Potently and selectively inhibits GABA reuptake
  • Exhibits neuroprotective properties
  • Protects neurons against mutant huntingtin-induced toxicity
  • Increases viability of SH-SY5Y cells
  • Inhibits GABA uptake in mouse GAT1 expressed in HEK293 cells
Catalog No. 50-004-07068 Supplier Medchemexpress LLC Supplier No. HYB0696A10MM/1ML
Tiagabine (hydrochloride) is an anticonvulsant agent and a selective inhibitor of the GAT-1 GABA transporter. It potently and selectively inhibits GABA reuptake and demonstrates neuroprotective properties.
  • Selective inhibitor of GAT-1 GABA transporter
  • Potently and selectively inhibits GABA reuptake
  • Exhibits neuroprotective properties
  • Protects neurons against mutant huntingtin-induced toxicity
  • Increases viability of SH-SY5Y cells
  • Inhibits GABA uptake in mouse GAT1 expressed in HEK293 cells