Learn More
Cayman Chemical TIagabInhydrochlorIde 10mg
SDP
Catalog No. 502995608
Encompass_Preferred
A GAT-1 inhibitor (IC50 = 49 nM); inhibits seizures induced by DMCM in mice (ED50 = 1.2 mg/kg, i.p.); reduces allodynia in a rodent model of neuropathic pain when used at a dose of 72.8 µmol/kg; acts synergistically with gabapentin to delay pain responses in mice in the hot plate test
Catalog No. 50-299-5608 Supplier Cayman Chemical Supplier No. 2292610 MG
A GAT-1 inhibitor (IC50 = 49 nM); inhibits seizures induced by DMCM in mice (ED50 = 1.2 mg/kg, i.p.); reduces allodynia in a rodent model of neuropathic pain when used at a dose of 72.8 µmol/kg; acts synergistically with gabapentin to delay pain responses in mice in the hot plate test