Learn More
Medchemexpress LLC Tiamulin | 55297-95-5 | 98.1% | 250 MG
SDP
Catalog No. 5000414340
Encompass_Preferred
Tiamulin is a diterpenic antibiotic widely used in pigs and poultry to control infectious diseases, particularly airsacculitis caused by Mycoplasma spp. This semisynthetic derivative of Pleuromutilin is highly effective against various Mycoplasma strains, Spirochaetes, and gram-positive bacteria by binding to rRNA and inhibiting protein production. It is less active against gram-negative bacteria. Tiamulin has been shown to be effective in treating avian intestinal spirochaetosis.
  • Diterpenic antibiotic for infectious diseases
  • Effective against Mycoplasma strains, Spirochaetes, and gram-positive bacteria
  • Semisynthetic derivative of Pleuromutilin
  • Inhibits protein production by binding to rRNA
  • Less active against gram-negative bacteria
  • Effective in treating avian intestinal spirochaetosis
Catalog No. 50-004-14340 Supplier Medchemexpress LLC Supplier No. 250MGHYB2060
Tiamulin is a diterpenic antibiotic widely used in pigs and poultry to control infectious diseases, particularly airsacculitis caused by Mycoplasma spp. This semisynthetic derivative of Pleuromutilin is highly effective against various Mycoplasma strains, Spirochaetes, and gram-positive bacteria by binding to rRNA and inhibiting protein production. It is less active against gram-negative bacteria. Tiamulin has been shown to be effective in treating avian intestinal spirochaetosis.
  • Diterpenic antibiotic for infectious diseases
  • Effective against Mycoplasma strains, Spirochaetes, and gram-positive bacteria
  • Semisynthetic derivative of Pleuromutilin
  • Inhibits protein production by binding to rRNA
  • Less active against gram-negative bacteria
  • Effective in treating avian intestinal spirochaetosis