Learn More
Medchemexpress LLC Tiamulin fumarate | 55297-96-6 | 99.3% | 1 ML
SDP
Catalog No. 5000410573
Encompass_Preferred
Tiamulin fumarate is a diterpenic antibiotic commonly used in pigs and poultry to control infectious diseases, including airsacculitis caused by Mycoplasma spp. This semisynthetic derivative of the diterpene antibiotic Pleuromutilin binds to rRNA, preventing protein production, and is highly effective against various Mycoplasma strains, Spirochaetes, and gram-positive bacteria.
  • Highly active against Mycoplasma strains (M. gallisepticum, M. synoviae, M. meleagridis, and M. iowae).
  • Effective against Spirochaetes (Brachyspira hyodysenteriae, Brachyspira innocens, B. pilosicoli, B. intermedia).
  • Active against gram-positive bacteria such as staphylococci, streptococci, Clostridia, and Arcanobacterium spp.
  • Mechanism of action involves binding to the peptidyl transferase slot on the ribosome, inhibiting protein synthesis.
  • Effective in treating avian intestinal spirochaetosis.
  • For research use only.
Catalog No. 50-004-10573 Supplier Medchemexpress LLC Supplier No. HYB2060A10MM/1ML
Tiamulin fumarate is a diterpenic antibiotic commonly used in pigs and poultry to control infectious diseases, including airsacculitis caused by Mycoplasma spp. This semisynthetic derivative of the diterpene antibiotic Pleuromutilin binds to rRNA, preventing protein production, and is highly effective against various Mycoplasma strains, Spirochaetes, and gram-positive bacteria.
  • Highly active against Mycoplasma strains (M. gallisepticum, M. synoviae, M. meleagridis, and M. iowae).
  • Effective against Spirochaetes (Brachyspira hyodysenteriae, Brachyspira innocens, B. pilosicoli, B. intermedia).
  • Active against gram-positive bacteria such as staphylococci, streptococci, Clostridia, and Arcanobacterium spp.
  • Mechanism of action involves binding to the peptidyl transferase slot on the ribosome, inhibiting protein synthesis.
  • Effective in treating avian intestinal spirochaetosis.
  • For research use only.