Learn More
Cayman Chemical Tiamulin (fumarate), Purity: ≥95%, CAS: 55297-96-6, 1 g
SDP
Catalog No. 502981202
Encompass_Preferred
A pleuromutilin antibiotic; active against various mycobacteria (MICs = 0.0025-0.25 µg/ml) and various Gram-positive bacteria, including MRSA; binds to the peptidyl transferase in the 50S ribosomal subunit to inhibit protein synthesis
Catalog No. 50-298-1202 Supplier Cayman Chemical Supplier No. 212161 G
A pleuromutilin antibiotic; active against various mycobacteria (MICs = 0.0025-0.25 µg/ml) and various Gram-positive bacteria, including MRSA; binds to the peptidyl transferase in the 50S ribosomal subunit to inhibit protein synthesis