Learn More
Tians Basic Protection Polypropylene Gowns
Catalog No. 191303615 Shop All Tians International Products
This item is not returnable. View return policy
Catalog No. 19-130-3615 Supplier Tians Supplier No. 8137812XL
Add to Cart
Add to Cart
This item is not returnable. View return policy
Low-lint polypropylene gowns for use in some critical environments, and many general application areas.
Ideal for environments where exposure to liquid contamination is not a concern.
  • Lightweight Full length
  • Elastic wrists
Specifications
Gender Unisex
For Use With (Application) Critical Environments, General Industrial
No. of Pockets None
Product Type Controlled Environments Gown
Packaging Type Cleanroom Packaged, 10 Pieces/Sealed Polybag, 5 Bags/Sealed Master Polybag
Closure Type Neck and Waist Ties
Disposable Single-use
Sterility Non-sterile
Latex Free Yes
Model 813781
Sleeve Style Long Sleeve
Certifications/Compliance ISO Cleanroom Class 7
Autoclavable Not Autoclavable