Learn More
Apexbio Technology LLC Tianeptine sodium(Synonyms: Stablon, Coaxil, Tatinol, Tianeurax, Salymbra, Tianeptine Sodium Salt), 10mM (in 1mL DMSO), CAS: 30123-17-2.
SDP
Catalog No. 5000060846
Encompass_Preferred
Tianeptine sodium (CAS 30123-17-2) is a small molecule known to selectively facilitate serotonin (5-HT) uptake in both in vitro and in vivo experimental models It exhibits minimal affinity toward various neurotransmitter receptors including serotonin and dopamine receptors (IC 10 M) and does not affect the uptake mechanisms of noradrenaline or dopamine Due to its distinct serotonergic modulation tianeptine sodium is extensively used in neuroscience research particularly in studies investigating the molecular basis and therapeutic potential of antidepressants
Catalog No. 50-000-60846 Supplier Apexbio Technology LLC Supplier No. A53225.1
Tianeptine sodium (CAS 30123-17-2) is a small molecule known to selectively facilitate serotonin (5-HT) uptake in both in vitro and in vivo experimental models It exhibits minimal affinity toward various neurotransmitter receptors including serotonin and dopamine receptors (IC 10 M) and does not affect the uptake mechanisms of noradrenaline or dopamine Due to its distinct serotonergic modulation tianeptine sodium is extensively used in neuroscience research particularly in studies investigating the molecular basis and therapeutic potential of antidepressants