Learn More
Abcam Tianeptine sodium, 5-HT uptake facilitator, 10MG
Catalog No. 502706141
Encompass_Preferred
MW 458.9 Da, Purity >99%. Selective 5-HT uptake facilitator. Shows antidepressant, analgesic, neuroprotective and cognitive enhancing effects in vivo. Orally active.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6141 Supplier Abcam Supplier No. AB14300210MG
MW 458.9 Da, Purity >99%. Selective 5-HT uptake facilitator. Shows antidepressant, analgesic, neuroprotective and cognitive enhancing effects in vivo. Orally active.
The product is subject to the following: Abcam Restricted Use Statement