Learn More
AdipoGen Tianeptine sodium salt
SDP
Catalog No. 502559949
Encompass_Preferred
Chemical. CAS 30123-17-2. Formula C21H24ClN2O4S . Na. MW 435.9 . 23.0. Synthetic. Tianeptine is an atypical antidepressant with diverse actions, most notably on the glutamatergic system. It is a selective facilitator of 5-HT serotonin uptake in vitro and in vivo. It attenuates glutamate release and modulates the function of the AMPA receptor. Tianeptine also acts as an agonist of the µ-opioid receptor EC50s = 194 and 641nM for human and mouse receptors, respectively. Might have anti-inflammatory properties through modulation of TLR and NLRP3 inflammasome signaling pathways.
Catalog No. 50-255-9949 Supplier AdipoGen Supplier No. CDXT0456M500
Chemical. CAS 30123-17-2. Formula C21H24ClN2O4S . Na. MW 435.9 . 23.0. Synthetic. Tianeptine is an atypical antidepressant with diverse actions, most notably on the glutamatergic system. It is a selective facilitator of 5-HT serotonin uptake in vitro and in vivo. It attenuates glutamate release and modulates the function of the AMPA receptor. Tianeptine also acts as an agonist of the µ-opioid receptor EC50s = 194 and 641nM for human and mouse receptors, respectively. Might have anti-inflammatory properties through modulation of TLR and NLRP3 inflammasome signaling pathways.