Learn More
Medchemexpress LLC Tianeptine sodium salt | 30123-17-2 | 99.7% | C21H24ClN2NaO4S | 50 MG
SDP
Catalog No. 5000433570
Encompass_Preferred
Tianeptine sodium salt is an atypical antidepressant that functions as a moderate-intensity agonist of the μ-opioid and δ-opioid receptors. It acts as a glutamate modulator, enhancing AMPA and antagonizing NMDA receptor activity. Providing neuroprotective, anti-inflammatory, and antioxidant effects, it helps alleviate depression and anxiety symptoms without causing sedation.
  • Atypical antidepressant
  • Moderate μ-opioid and δ-opioid receptor agonist
  • Glutamate modulator (enhances AMPA, antagonizes NMDA)
  • Neuroprotective, anti-inflammatory, and antioxidant
  • Alleviates depression and anxiety without sedation
  • Low cardiotoxicity and minimal drug-drug interactions
Catalog No. 50-004-33570 Supplier Medchemexpress LLC Supplier No. HY90003A50MG
Tianeptine sodium salt is an atypical antidepressant that functions as a moderate-intensity agonist of the μ-opioid and δ-opioid receptors. It acts as a glutamate modulator, enhancing AMPA and antagonizing NMDA receptor activity. Providing neuroprotective, anti-inflammatory, and antioxidant effects, it helps alleviate depression and anxiety symptoms without causing sedation.
  • Atypical antidepressant
  • Moderate μ-opioid and δ-opioid receptor agonist
  • Glutamate modulator (enhances AMPA, antagonizes NMDA)
  • Neuroprotective, anti-inflammatory, and antioxidant
  • Alleviates depression and anxiety without sedation
  • Low cardiotoxicity and minimal drug-drug interactions