Learn More
Medchemexpress LLC Tianeptine (sodium salt) | 30123-17-2 | 99.9% | C21H24ClN2NaO4S | 1 ML
SDP
Catalog No. 5000433112
Encompass_Preferred
Tianeptine sodium salt, with chemical formula C21H24ClN2NaO4S and CAS number 30123-17-2, is provided as a 10 mM solution in 1 mL of DMSO. This ready-to-use format is designed for various research applications, including both in vitro and in vivo studies.
  • Ready-to-use 10 mM solution in DMSO
  • Ideal for in vitro and in vivo research applications
  • Recommended storage at -80°C for 6 months or -20°C for 1 month in solvent
  • High chemical purity of the active compound
Catalog No. 50-004-33112 Supplier Medchemexpress LLC Supplier No. HY90003A10MM/1ML
Tianeptine sodium salt, with chemical formula C21H24ClN2NaO4S and CAS number 30123-17-2, is provided as a 10 mM solution in 1 mL of DMSO. This ready-to-use format is designed for various research applications, including both in vitro and in vivo studies.
  • Ready-to-use 10 mM solution in DMSO
  • Ideal for in vitro and in vivo research applications
  • Recommended storage at -80°C for 6 months or -20°C for 1 month in solvent
  • High chemical purity of the active compound