Learn More
Medchemexpress LLC Tianeptine sodium salt | 30123-17-2 | 99.9% | C21H24ClN2NaO4S | 100 MG
SDP
Catalog No. 5000433367
Encompass_Preferred
Tianeptine sodium salt is an atypical antidepressant and a moderate μ-opioid and δ-opioid receptor agonist. It acts as a glutamate modulator, enhancing AMPA receptors and antagonizing NMDA receptors. The compound increases α1 adrenergic receptor sensitivity and offers neuroprotective, anti-inflammatory, and antioxidant effects under stress/inflammation. It inhibits MMP-9 by suppressing the PI3K/Akt-mediated NF-κB pathway, helping to alleviate depression and anxiety without sedative effects.
  • Moderate μ-opioid and δ-opioid receptor agonist.
  • Modulates glutamate receptors (AMPA enhancer, NMDA antagonist).
  • Boosts α1 adrenergic receptor sensitivity.
  • Offers neuroprotective, anti-inflammatory, and antioxidant benefits.
  • Inhibits MMP-9 via PI3K/Akt-mediated NF-κB pathway.
  • Reduces depression and anxiety without sedation.
  • Enhances cognition and memory.
  • Provides analgesic effects.
Catalog No. 50-004-33367 Supplier Medchemexpress LLC Supplier No. HY90003A100MG
Tianeptine sodium salt is an atypical antidepressant and a moderate μ-opioid and δ-opioid receptor agonist. It acts as a glutamate modulator, enhancing AMPA receptors and antagonizing NMDA receptors. The compound increases α1 adrenergic receptor sensitivity and offers neuroprotective, anti-inflammatory, and antioxidant effects under stress/inflammation. It inhibits MMP-9 by suppressing the PI3K/Akt-mediated NF-κB pathway, helping to alleviate depression and anxiety without sedative effects.
  • Moderate μ-opioid and δ-opioid receptor agonist.
  • Modulates glutamate receptors (AMPA enhancer, NMDA antagonist).
  • Boosts α1 adrenergic receptor sensitivity.
  • Offers neuroprotective, anti-inflammatory, and antioxidant benefits.
  • Inhibits MMP-9 via PI3K/Akt-mediated NF-κB pathway.
  • Reduces depression and anxiety without sedation.
  • Enhances cognition and memory.
  • Provides analgesic effects.