Description
- Material: Environstar™ SPP
- Breathable
- Elastic wrist
- Three pockets
- Color: White
Specifications
Specifications
| Product Type | Controlled Environments Lab Coat |
| Color | White |
| Closure Type | Snaps |
| Cuff Style | Elastic |
| Disposable | Single-use |
| Sterility | Non-sterile |
| Weight (English) | 10.12 lb. |
| Cleanroom Class | Class 10,000 (ISO 7) |