Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tians Epic™ High Performance Static Dissipative Cleanroom Sleeves

Catalog No. 19138552 Shop All Tians International Products
Change view
Click to view available options
Cleanroom Class:
Class 1000 (ISO 6)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Cleanroom Class
19-138-552 Class 1000 (ISO 6)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-138-552 Supplier Tians Supplier No. 8268318
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Lint free polyethylene and polypropylene sleeves for use in critical, ESD and low particulate clean environments.

  • Features elastic trimmed ends and tapered length for proper fit
  • Suitable for exposure to bloodborne pathogens, non-hazardous liquids and any wet general maintenance areas
  • Non-linting Environstar-MP fabric is constructed from unique laminated and breathable membrane
  • ISO Class 6 Compatible/Category 2 Garment
  • White in color
  • Length: 18 in.

Specifications

Product Type Controlled Environment Sleeve
Material Polyethylene-coated Polypropylene
Color White
Sterility Non-sterile
Disposable Single-use
Closure Type Elastic Ends
Seam Type Serged
Latex Free Yes
Model 82683-18
Length (English) 18 in.
Length (Metric) 45.7 cm
Cleanroom Class Class 1000 (ISO 6)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.