Learn More
Tians Epic™ Premium Coated Barrier Cleanroom Gowns
Catalog No. 19150352 Shop All Tians International Products
Change view
Click to view available options
Disposable:
Single-use
Catalog No. Disposable
19-150-352 Single-use
1 options
This item is not returnable. View return policy
Catalog No. 19-150-352 Supplier Tians Supplier No. 816851TSM
Add to Cart
Add to Cart
This item is not returnable. View return policy
Static dissipative, lint-free film coated gown for use in critical and low particulate ISO cleanroom class 6 environments.
  • Latex free, lint-free film coated gown for use in critical and low particulate clean environments
  • Static dissipative
  • Fabric is suitable for exposure to bloodborne pathogens, some hazardous liquids, and where liquid splash is a concern
  • Elastic wrists, elastic thumb loop, wrap-around neck ties, wrap-around waist ties
  • Serged seam construction
  • Fabric meets or exceeds NIOSH, OSHA, ASHP and ONS guidelines for exposure to chemotherapy drugs
  • Fabric is constructed from a blend of polyethylene and polypropylene into a unique breathable membrane
  • ISO Cleanroom Class 6
  • Helmke Drum Testing IEST-RP-CC003.3
Specifications
Product Type Controlled Environments Gown
Gender Unisex
No. of Pockets None
Disposable Single-use
Sterility Non-sterile
Autoclavable Not Autoclavable