Learn More
Tians epic™ Lab Coats
Catalog No. p-3649498 Shop All Tians International Products
Change view
Click to view available options
Color:
White
No. of Pockets:
1
Size:
Medium
Large
X-Large
2X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color No. of Pockets Size
19-130-1944E White 1 2X-Large
19-130-1944C White 1 Large
19-130-1944B White 1 Medium
19-130-1944D White 1 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-130-1944E Supplier Tians Supplier No. 8458812XL
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Specifications
Product Type Controlled Environments Lab Coat
Size 2X-Large
Material Spunbond Polypropylene
Color White
Closure Type Snaps
Cuff Style Elastic
No. of Pockets 1
Collar Type Folded
Latex Free Yes
Packaging Type 10 pieces per inner bag, 5 inner bags per master bag, 50 pieces per case
Cleanroom Class Class 10,000 (ISO 7)