Learn More
Apexbio Technology LLC Tiaprofenic acid 33005-95-7 25mg
SDP
Catalog No. 5000093769
Encompass_Preferred
Tiaprofenic acid (CAS 33005-95-7) is a non-steroidal anti-inflammatory compound of the arylpropionic acid class It exerts its pharmacological effect primarily by inhibiting cyclooxygenase (COX) isoenzymes resulting in reduced synthesis of prostaglandins involved in inflammation and pain signaling Widely studied for its application in models of inflammatory disease tiaprofenic acid is utilized in biomedical research to investigate mechanisms underlying arthritis pain and to evaluate the modulation of prostanoid-mediated inflammatory pathways
Catalog No. 50-000-93769 Supplier Apexbio Technology LLC Supplier No. A469925
Add to Cart
Add to Cart
Tiaprofenic acid (CAS 33005-95-7) is a non-steroidal anti-inflammatory compound of the arylpropionic acid class It exerts its pharmacological effect primarily by inhibiting cyclooxygenase (COX) isoenzymes resulting in reduced synthesis of prostaglandins involved in inflammation and pain signaling Widely studied for its application in models of inflammatory disease tiaprofenic acid is utilized in biomedical research to investigate mechanisms underlying arthritis pain and to evaluate the modulation of prostanoid-mediated inflammatory pathways