Learn More
Medchemexpress LLC Tiaprofenic acid-d3 | 2740831-64-3 | 96.3% | 263.33 | C14H9D3O3S | 100 UG
SDP
Catalog No. 5000240967
Encompass_Preferred
Tiaprofenic acid-d3 is a deuterium-labeled analogue of tiaprofenic acid intended for use as a research reagent and analytical standard in studies of cyclooxygenase (COX) pathways. It is provided as an isotopically labeled, high-purity compound suitable for biochemical assays and mass spectrometry applications.
  • Deuterium-labeled compound for isotopic tracing and internal standard use.
  • High purity for reliable analytical performance.
  • Available in small, pre-weighed quantities for convenient handling.
  • Compatible with mass spectrometry and chromatographic methods.
  • Suitable for metabolic studies and quantitative assays.
Catalog No. 50-002-40967 Supplier Medchemexpress LLC Supplier No. HY106579S100UG
Add to Cart
Add to Cart
Tiaprofenic acid-d3 is a deuterium-labeled analogue of tiaprofenic acid intended for use as a research reagent and analytical standard in studies of cyclooxygenase (COX) pathways. It is provided as an isotopically labeled, high-purity compound suitable for biochemical assays and mass spectrometry applications.
  • Deuterium-labeled compound for isotopic tracing and internal standard use.
  • High purity for reliable analytical performance.
  • Available in small, pre-weighed quantities for convenient handling.
  • Compatible with mass spectrometry and chromatographic methods.
  • Suitable for metabolic studies and quantitative assays.