Learn More
Medchemexpress LLC Tiaprofenic acid-d3 | 2740831-64-3 | 96.3% | 263.33 g/mol | C14H9D3O3S | 1 MG
SDP
Supplier:  Medchemexpress LLC HY106579S1MG
Encompass_Preferred
Tiaprofenic acid-d3 is a deuterium-labeled analogue of the nonsteroidal anti-inflammatory compound tiaprofenic acid, provided for research use only. It serves as an isotopically labeled internal standard for analytical workflows and supports studies of drug metabolism, pharmacokinetics, and quantitative bioanalysis.
  • Deuterium-labeled analogue for isotopic tracing.
  • Suitable as an internal standard for LC-MS/MS assays.
  • Supports drug metabolism and pharmacokinetic studies.
  • High purity for analytical applications (96.3% reported).
  • Supplied in small mg quantities for laboratory research.
Catalog No. 50-002-41012