Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tiaprofenic acid-d3 | 2740831-64-3 | 96.3% | 263.33 g/mol | C14H9D3O3S | 5 MG
SDP

Catalog No. 5000240302
Encompass_Preferred

Tiaprofenic acid-d3 is a deuterium-labeled analogue of tiaprofenic acid provided as a research-grade chemical standard. It is intended for use as an internal standard or tracer in analytical, pharmacokinetic, and mass spectrometry studies of the nonsteroidal anti-inflammatory compound, and is supplied in small, accurately weighed quantities for laboratory applications.

  • Deuterium-labeled analog for use as an internal standard.
  • Purity 96.3%.
  • Molecular formula C14H9D3O3S.
  • Molecular weight 263.33 g/mol.
  • Available in small pack sizes such as 5 mg.
  • Supplied as a solid for analytical and tracer applications.
  • For research use only, not for human or clinical use.

Catalog No. 50-002-40302 Supplier Medchemexpress LLC Supplier No. HY106579S5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tiaprofenic acid-d3 is a deuterium-labeled analogue of tiaprofenic acid provided as a research-grade chemical standard. It is intended for use as an internal standard or tracer in analytical, pharmacokinetic, and mass spectrometry studies of the nonsteroidal anti-inflammatory compound, and is supplied in small, accurately weighed quantities for laboratory applications.

  • Deuterium-labeled analog for use as an internal standard.
  • Purity 96.3%.
  • Molecular formula C14H9D3O3S.
  • Molecular weight 263.33 g/mol.
  • Available in small pack sizes such as 5 mg.
  • Supplied as a solid for analytical and tracer applications.
  • For research use only, not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.