Learn More
Medchemexpress LLC Tiaramide hydrochloride | 35941-71-0 | MFCD01702832 | 98.7% | 392.30 | C15H19Cl2N3O3S | 5 MG
SDP
Catalog No. 5000262284
Encompass_Preferred
Tiaramide hydrochloride is the hydrochloride salt of tiaramide, an analgesic research compound provided in milligram quantities for laboratory use. It is a light yellow solid with molecular formula C15H19Cl2N3O3S and molecular weight 392.30 g·mol-1. Typical purity is 98.7%, and the compound is commonly handled in DMSO for in vitro and pharmacological studies.
  • Hydrochloride salt form for improved stability
  • High purity (approximately 98.7%)
  • Available in milligram-scale sizes for research use
  • Light yellow solid suitable for analytical testing
  • Soluble in DMSO for biological assays
Catalog No. 50-002-62284 Supplier Medchemexpress LLC Supplier No. HY134055MG
Tiaramide hydrochloride is the hydrochloride salt of tiaramide, an analgesic research compound provided in milligram quantities for laboratory use. It is a light yellow solid with molecular formula C15H19Cl2N3O3S and molecular weight 392.30 g·mol-1. Typical purity is 98.7%, and the compound is commonly handled in DMSO for in vitro and pharmacological studies.
  • Hydrochloride salt form for improved stability
  • High purity (approximately 98.7%)
  • Available in milligram-scale sizes for research use
  • Light yellow solid suitable for analytical testing
  • Soluble in DMSO for biological assays