Learn More
Medchemexpress LLC Tibenelast sodium | 105102-18-9 | MFCD00864802 | >98.0% | 288.29 g·mol⁻¹ | C13H13NaO4S | 25 MG
SDP
Catalog No. 5000249017
Encompass_Preferred
Tibenelast sodium is a small-molecule phosphodiesterase inhibitor supplied as the sodium salt for preclinical research. It is used to modulate cyclic nucleotide signaling in biochemical and cellular studies and is typically provided as a white to off-white solid suitable for dissolution in common organic solvents.
  • Phosphodiesterase inhibition activity for PDE-related studies.
  • Suitable for biochemical and cellular assays.
  • Provided as a stable sodium salt to aid solubility in DMSO and other solvents.
  • Commonly available in small research pack sizes for lab-scale experiments.
  • Chemical identity confirmed (CAS 105102-18-9); consult the certificate of analysis for lot-specific purity.
Catalog No. 50-002-49017 Supplier Medchemexpress LLC Supplier No. HY10170525MG
Tibenelast sodium is a small-molecule phosphodiesterase inhibitor supplied as the sodium salt for preclinical research. It is used to modulate cyclic nucleotide signaling in biochemical and cellular studies and is typically provided as a white to off-white solid suitable for dissolution in common organic solvents.
  • Phosphodiesterase inhibition activity for PDE-related studies.
  • Suitable for biochemical and cellular assays.
  • Provided as a stable sodium salt to aid solubility in DMSO and other solvents.
  • Commonly available in small research pack sizes for lab-scale experiments.
  • Chemical identity confirmed (CAS 105102-18-9); consult the certificate of analysis for lot-specific purity.