Learn More
Apexbio Technology LLC TIC10 41276-02-2 10mM (in 1mL DMSO)
SDP
Catalog No. 5000061496
Encompass_Preferred
TIC10 (TRAIL-inducing compound 10) is a small molecule inhibitor with oral bioavailability capable of penetrating the blood-brain barrier It functions primarily by simultaneously inhibiting Akt and ERK pathways resulting in Foxo3a nuclear translocation and transcriptional activation of TRAIL (tumor necrosis factor-related apoptosis-inducing ligand) TRAIL is involved in apoptosis induction in cancer cells often compromised during tumor progression TIC10 triggers the expression of TRAIL independently of p53 status enabling apoptosis in diverse cancer cell types In preclinical xenograft models and orthotopic glioblastoma studies TIC10 has demonstrated TRAIL-mediated antitumor activity supporting its use in cancer biology research and therapeutic investigations
Catalog No. 50-000-61496 Supplier Apexbio Technology LLC Supplier No. A86195.1
TIC10 (TRAIL-inducing compound 10) is a small molecule inhibitor with oral bioavailability capable of penetrating the blood-brain barrier It functions primarily by simultaneously inhibiting Akt and ERK pathways resulting in Foxo3a nuclear translocation and transcriptional activation of TRAIL (tumor necrosis factor-related apoptosis-inducing ligand) TRAIL is involved in apoptosis induction in cancer cells often compromised during tumor progression TIC10 triggers the expression of TRAIL independently of p53 status enabling apoptosis in diverse cancer cell types In preclinical xenograft models and orthotopic glioblastoma studies TIC10 has demonstrated TRAIL-mediated antitumor activity supporting its use in cancer biology research and therapeutic investigations