Learn More
Apexbio Technology LLC TIC10, 20mg. Cas: 41276-02-2 MFCD: MFCD25976764. Potent Akt/ERK inhibitor
SDP
Supplier:  Apexbio Technology LLC A861920
Encompass_Preferred
TIC10 (TRAIL-inducing compound 10) is a potent and stable small molecule that is orally active. It induces TRAIL transcriptionally independent of p53 and crosses the blood-brain barrier. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5166