Learn More
Apexbio Technology LLC Ticagrelor 274693-27-5 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062430
Encompass_Preferred
Ticagrelor (CAS 274693-27-5) is an orally active reversible antagonist targeting the platelet P2Y12 receptor By competitively binding to P2Y12 ticagrelor inhibits ADP-mediated platelet aggregation resulting in dose-dependent suppression of platelet activation and thrombosis in vitro Unlike other P2Y12 inhibitors requiring metabolic activation ticagrelor acts without metabolic transformation though it is primarily metabolized by CYP3A4 and CYP2C19 enzymes This compound is frequently employed in biomedical research exploring platelet function thrombosis mechanisms and drug-drug interactions involving cytochrome P450 enzymes
Catalog No. 50-000-62430 Supplier Apexbio Technology LLC Supplier No. B21665.1
Ticagrelor (CAS 274693-27-5) is an orally active reversible antagonist targeting the platelet P2Y12 receptor By competitively binding to P2Y12 ticagrelor inhibits ADP-mediated platelet aggregation resulting in dose-dependent suppression of platelet activation and thrombosis in vitro Unlike other P2Y12 inhibitors requiring metabolic activation ticagrelor acts without metabolic transformation though it is primarily metabolized by CYP3A4 and CYP2C19 enzymes This compound is frequently employed in biomedical research exploring platelet function thrombosis mechanisms and drug-drug interactions involving cytochrome P450 enzymes