Learn More
Medchemexpress LLC Ticagrelor impurity 10 | 98.5% | 562.63 g·mol⁻¹ | C26H32F2N6O4S | 10 MG
SDP
Catalog No. 5000298381
Encompass_Preferred
Ticagrelor impurity 10 is a research-grade impurity of the antiplatelet agent ticagrelor, supplied for analytical reference, impurity profiling, and method development. The material is provided at high stated purity (98.51%) and is characterized by a molecular weight of 562.63 g·mol⁻¹. Small-mass packages are offered for laboratory use, and accompanying documentation (SDS, COA) is available.
  • Research-grade impurity for analytical and QC applications.
  • High stated purity (98.51%) for reliable reference standards.
  • Available in small milligram quantities suitable for method development.
  • Characterized molecular weight for analytical confirmation.
  • SDS and COA available to support safe handling and verification.
Catalog No. 50-002-98381 Supplier Medchemexpress LLC Supplier No. HY77732A10MG
Ticagrelor impurity 10 is a research-grade impurity of the antiplatelet agent ticagrelor, supplied for analytical reference, impurity profiling, and method development. The material is provided at high stated purity (98.51%) and is characterized by a molecular weight of 562.63 g·mol⁻¹. Small-mass packages are offered for laboratory use, and accompanying documentation (SDS, COA) is available.
  • Research-grade impurity for analytical and QC applications.
  • High stated purity (98.51%) for reliable reference standards.
  • Available in small milligram quantities suitable for method development.
  • Characterized molecular weight for analytical confirmation.
  • SDS and COA available to support safe handling and verification.