Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ticagrelor impurity 11 | 220352-36-3 | ≥95.0% | C10H8F2O2 | 5 G
SDP

Catalog No. 5000434919
Encompass_Preferred

Ticagrelor impurity 11 is an impurity of Ticagrelor. It appears as a solid with a molecular weight of 198.17. This product is for research use only.

  • Purity: ≥95.0%
  • Storage as powder: -20 °C for 3 years, 4 °C for 2 years
  • Storage in solvent: -80 °C for 6 months, -20 °C for 1 month

Catalog No. 50-004-34919 Supplier Medchemexpress LLC Supplier No. HY779645G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ticagrelor impurity 11 is an impurity of Ticagrelor. It appears as a solid with a molecular weight of 198.17. This product is for research use only.

  • Purity: ≥95.0%
  • Storage as powder: -20 °C for 3 years, 4 °C for 2 years
  • Storage in solvent: -80 °C for 6 months, -20 °C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.