Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ticagrelor impurity 11 | 220352-36-3 | ≥95.0% | C10H8F2O2 | 500 MG
SDP

Catalog No. 5000434444
Encompass_Preferred

Ticagrelor impurity 11 is an impurity of Ticagrelor, presented as a solid powder. This chemical is intended for laboratory research use and the manufacture of substances.

  • Impurity of Ticagrelor
  • Presented as a solid powder
  • For laboratory research use only
  • Purity greater than or equal to 95.0%
  • Stable as powder at -20°C for 3 years or 4°C for 2 years
  • Stable in solvent at -80°C for 6 months or -20°C for 1 month

Catalog No. 50-004-34444 Supplier Medchemexpress LLC Supplier No. HY77964500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ticagrelor impurity 11 is an impurity of Ticagrelor, presented as a solid powder. This chemical is intended for laboratory research use and the manufacture of substances.

  • Impurity of Ticagrelor
  • Presented as a solid powder
  • For laboratory research use only
  • Purity greater than or equal to 95.0%
  • Stable as powder at -20°C for 3 years or 4°C for 2 years
  • Stable in solvent at -80°C for 6 months or -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.