Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ticagrelor impurity 16 | 2096989-56-7 | 99.5% | 522.57 | C23H28F2N6O4S | 10 MG
SDP

Catalog No. 5000298694
Encompass_Preferred

Ticagrelor impurity 16 is a synthetic impurity and analytical reference for the antiplatelet drug ticagrelor, supplied for research and analytical applications such as impurity profiling and method development. Certificate of analysis and safety data sheet are available for batch-specific information.

  • Suitable for analytical reference and impurity profiling.
  • CAS number: 2096989-56-7.
  • Molecular formula: C23H28F2N6O4S; molecular weight: 522.57.
  • Typical purity: 99.5% (HPLC) as indicated on batch COA.
  • Available in small quantities (1 mg-100 mg), including 10 MG.
  • Storage: room temperature (up to 3 years); in solvent: -80°C (2 years) or -20°C (1 year).

Catalog No. 50-002-98694 Supplier Medchemexpress LLC Supplier No. HY7849510MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ticagrelor impurity 16 is a synthetic impurity and analytical reference for the antiplatelet drug ticagrelor, supplied for research and analytical applications such as impurity profiling and method development. Certificate of analysis and safety data sheet are available for batch-specific information.

  • Suitable for analytical reference and impurity profiling.
  • CAS number: 2096989-56-7.
  • Molecular formula: C23H28F2N6O4S; molecular weight: 522.57.
  • Typical purity: 99.5% (HPLC) as indicated on batch COA.
  • Available in small quantities (1 mg-100 mg), including 10 MG.
  • Storage: room temperature (up to 3 years); in solvent: -80°C (2 years) or -20°C (1 year).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.