Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ticagrelor impurity 25 | 1616703-93-5 | 96.9% | C25H30F2N6O5S | 100 MG
SDP

Catalog No. 5000432088
Encompass_Preferred

Ticagrelor impurity 25 is an impurity of Ticagrelor. It is for research use only.

  • Molecular weight: 564.60
  • Appearance: Solid
  • Shipping at room temperature in continental US; may vary elsewhere
  • Store at 4°C, protect from light
  • Store in solvent at -80°C for 6 months or -20°C for 1 month, protecting from light

Catalog No. 50-004-32088 Supplier Medchemexpress LLC Supplier No. HY78999100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ticagrelor impurity 25 is an impurity of Ticagrelor. It is for research use only.

  • Molecular weight: 564.60
  • Appearance: Solid
  • Shipping at room temperature in continental US; may vary elsewhere
  • Store at 4°C, protect from light
  • Store in solvent at -80°C for 6 months or -20°C for 1 month, protecting from light

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.