Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ticagrelor impurity 27 | 2096989-55-6 | 98.9% | 522.57 g/mol | C23H28F2N6O4S | 10 MG
SDP

Catalog No. 5000298606
Encompass_Preferred

Ticagrelor impurity 27 is an analytical standard and characterized drug intermediate supplied for research and analytical testing related to ticagrelor. The material is provided as a high-purity solid with documented molecular formula C23H28F2N6O4S and molecular weight 522.57 g/mol; accompanying safety and quality documents are available.

  • High purity: 98.9%.
  • Molecular formula C23H28F2N6O4S.
  • Molecular weight 522.57 g/mol.
  • Available in small pack sizes including 1 mg, 5 mg, 10 mg, 50 mg, and 100 mg.
  • Intended for research and analytical use only; not for human consumption.
  • Comes with COA and SDS for quality and safety information.

Catalog No. 50-002-98606 Supplier Medchemexpress LLC Supplier No. HY7849610MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ticagrelor impurity 27 is an analytical standard and characterized drug intermediate supplied for research and analytical testing related to ticagrelor. The material is provided as a high-purity solid with documented molecular formula C23H28F2N6O4S and molecular weight 522.57 g/mol; accompanying safety and quality documents are available.

  • High purity: 98.9%.
  • Molecular formula C23H28F2N6O4S.
  • Molecular weight 522.57 g/mol.
  • Available in small pack sizes including 1 mg, 5 mg, 10 mg, 50 mg, and 100 mg.
  • Intended for research and analytical use only; not for human consumption.
  • Comes with COA and SDS for quality and safety information.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.