Learn More
Medchemexpress LLC Ticagrelor impurity 27 | 2096989-55-6 | 99.0% | C23H28F2N6O4S | 5 MG
SDP
Catalog No. 5000433290
Encompass_Preferred
Ticagrelor impurity 27 is an impurity of Ticagrelor (HY-10064).
  • High purity (99.01%)
  • Solid appearance
  • Molecular weight of 522.57
  • Store powder at -20°C for 3 years, 4°C for 2 years
  • Store in solvent at -80°C for 6 months, -20°C for 1 month
  • For research use only
Catalog No. 50-004-33290 Supplier Medchemexpress LLC Supplier No. HY784965MG
Ticagrelor impurity 27 is an impurity of Ticagrelor (HY-10064).
  • High purity (99.01%)
  • Solid appearance
  • Molecular weight of 522.57
  • Store powder at -20°C for 3 years, 4°C for 2 years
  • Store in solvent at -80°C for 6 months, -20°C for 1 month
  • For research use only