Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ticagrelor impurity 5 | 1863036-49-0 | >99.7% | C₂₆H₃₂F₂N₆O₅S | 1 MG
SDP

Catalog No. 5000433249
Encompass_Preferred

Ticagrelor impurity 5 is an impurity of Ticagrelor (HY-10064) with a molecular weight of 578.63 and a purity of >99.66%. It appears as an off-white to yellow solid and is stable under recommended storage conditions.

  • Impurity of Ticagrelor
  • Chemical formula C₂₆H₃₂F₂N₆O₅S
  • CAS number 1863036-49-0
  • Appearance as an off-white to yellow solid
  • Stable under recommended storage conditions
  • Store at 4°C, stored under nitrogen
  • In solvent, store at -80°C for 6 months or -20°C for 1 month, stored under nitrogen
  • Shipping at room temperature if less than 2 weeks

Catalog No. 50-004-33249 Supplier Medchemexpress LLC Supplier No. HY785461MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ticagrelor impurity 5 is an impurity of Ticagrelor (HY-10064) with a molecular weight of 578.63 and a purity of >99.66%. It appears as an off-white to yellow solid and is stable under recommended storage conditions.

  • Impurity of Ticagrelor
  • Chemical formula C₂₆H₃₂F₂N₆O₅S
  • CAS number 1863036-49-0
  • Appearance as an off-white to yellow solid
  • Stable under recommended storage conditions
  • Store at 4°C, stored under nitrogen
  • In solvent, store at -80°C for 6 months or -20°C for 1 month, stored under nitrogen
  • Shipping at room temperature if less than 2 weeks

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.