Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ticagrelor impurity 8 | 1863036-51-4 | 99.0% | 594.63 | C26H32F2N6O6S | 10 MG
SDP

Catalog No. 5000298394
Encompass_Preferred

Ticagrelor impurity 8 is an identified impurity of the antiplatelet drug ticagrelor, provided as a research-grade analytical standard for impurity profiling, method development, and compound characterization. It is intended for laboratory research purposes and not for human or clinical use.

  • CAS number: 1863036-51-4.
  • Purity: 98.96% (as reported).
  • Molecular formula: C26H32F2N6O6S.
  • Molecular weight: 594.63 g/mol.
  • Available package sizes: 1 mg, 5 mg, 10 mg, 50 mg, 100 mg.

Catalog No. 50-002-98394 Supplier Medchemexpress LLC Supplier No. HY7854710MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ticagrelor impurity 8 is an identified impurity of the antiplatelet drug ticagrelor, provided as a research-grade analytical standard for impurity profiling, method development, and compound characterization. It is intended for laboratory research purposes and not for human or clinical use.

  • CAS number: 1863036-51-4.
  • Purity: 98.96% (as reported).
  • Molecular formula: C26H32F2N6O6S.
  • Molecular weight: 594.63 g/mol.
  • Available package sizes: 1 mg, 5 mg, 10 mg, 50 mg, 100 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.