Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ticagrelor impurity 8 | 1863036-51-4 | 99.0% | C26H32F2N6O6S | 5 MG
SDP

Catalog No. 5000433325
Encompass_Preferred

Ticagrelor impurity 8 is a solid chemical compound with the formula C26H32F2N6O6S. It is intended for research use only as a laboratory chemical and in the manufacture of substances.

  • Purity of 98.96% (HPLC)
  • Stable under recommended storage conditions
  • Appearance: Light yellow to yellow solid
  • Store at room temperature for 3 years, or in solvent at -80°C for 2 years, or -20°C for 1 year
  • Harmful if swallowed
  • Causes skin and serious eye irritation
  • May cause respiratory irritation

Catalog No. 50-004-33325 Supplier Medchemexpress LLC Supplier No. HY785475MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ticagrelor impurity 8 is a solid chemical compound with the formula C26H32F2N6O6S. It is intended for research use only as a laboratory chemical and in the manufacture of substances.

  • Purity of 98.96% (HPLC)
  • Stable under recommended storage conditions
  • Appearance: Light yellow to yellow solid
  • Store at room temperature for 3 years, or in solvent at -80°C for 2 years, or -20°C for 1 year
  • Harmful if swallowed
  • Causes skin and serious eye irritation
  • May cause respiratory irritation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.