Learn More
Abcam Ticagrelor, P2Y12 receptor antagonist, 25MG
Catalog No. 502672651
Encompass_Preferred
MW 522.6 Da, Purity >98%. Reversible antagonist of the platelet purinergic P2Y12 receptor, the main receptor responsible for ADP-induced platelet aggregation. Changes the conformation of the P2Y12 receptor, resulting in reversible, concentration dependent inhibition of the receptor.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-2651 Supplier Abcam Supplier No. AB23036625MG
MW 522.6 Da, Purity >98%. Reversible antagonist of the platelet purinergic P2Y12 receptor, the main receptor responsible for ADP-induced platelet aggregation. Changes the conformation of the P2Y12 receptor, resulting in reversible, concentration dependent inhibition of the receptor.
The product is subject to the following: Abcam Restricted Use Statement